Personal Cleaning Station, Dry Recovery-Only, 2D-1 (PCS-WM) HEPA ORDLOC 652
Supplier: Tiger-Vac International
113386A
77008-234EA 1194.64 USD
77008-234
Personal Cleaning Station, Dry Recovery-Only, 2D-1 (PCS-WM) HEPA ORDLOC 652
Vacuum Cleaners
HEPA filtered 'personal cleaning station' with small footprint intended to be wall mounted.
  • Complies with NFPA 652 paragraph 8.4.2.2.1.7 and NFPA 654 paragraph 8.4.3.7 (1) to (7)
  • Protected against ingress of dust
  • PCS 'Personal Cleaning Station' with small footprint intended to be wall mounted
  • Ideal for locker rooms, changing rooms and gowning rooms
  • Employees can remove (vacuum) dust and powder from their clothing before or after their shift
  • Can also be removed from wall and used as a portable HEPA vac
  • Heavy duty two-stage by-pass motor
  • For dry recovery only
  • Stainless steel tank Type 430 fits any application
  • Includes a static dissipative suction hose and round brush
  • Other static dissipative accessories are available as optional
HEPA filter with an efficiency of 99.995% at 0.3 micron. Tested IEST-RP-CC001. H14 by MPPS method as per EN 1822 and OSHA compliant. All of our HEPA vacuum systems are aerosol leak tested before leaving our facility - included.
Suction inlet: 1.5"
Certifications: ANSI/UL 1017. CSA C22.2 No. 243.
Ordering information: For available options or accessories; contact your Avantor representative.