Vacuum Cleaner, Dry Recovery-Only, C-10 EX (4W)
Supplier: Tiger-Vac International
111805A
77008-228EA 3146.56 USD
77008-228
Vacuum Cleaner, Dry Recovery-Only, C-10 EX (4W)
Vacuum Cleaners
HEPA filtered vacuum used to recover combustible/conductive dusts in Hazardous Locations Division 2. Excellent seller in addititve manufacturing and 3D printing.
  • Ideal for housekeeping and general cleaning
  • Includes four HEPA filters, Class H14, individually factory tested
  • Heavy duty two-stage by-pass motor
  • Longer life brushless motor available; always with 4×H14 HEPA filters
  • Material to be recovered within a conductive cloth recovery bag
  • Includes an indicator light to warn of vacuum blockage
  • Includes a complete static dissipative tool kit
Recovery tank made of stainless steel Type 430 (recovery tank made of stainless steel Type 304 is also available).
Suction inlet: 1.5"
Certifications: Dust ignition protected. Cleaning machines for Hazardous Locations (Classes C1618 01 & C1618 81). Division 2, Class II, Groups F & G.
Ordering information: For available options or accessories; contact your Avantor representative.