Vacuum Cleaner, AS-400 MR HEPA (W + D)
Supplier: Tiger-Vac International
110776A
77008-226EA 1089.65 USD
77008-226
Vacuum Cleaner, AS-400 MR HEPA (W + D)
Vacuum Cleaners
HEPA filtered vacuum for wet and dry recovery, designed for the recovery of asbestos, lead, mold, etc.
  • Designed for the recovery of asbestos, lead, mold, etc.
  • Lightweight and portable
  • Heavy duty two-stage by-pass motor
  • Equipped with a three-stage filtration system
  • Middle ring for wet and dry recovery - included
  • Heavy duty plastic recovery tank
  • Stainless steel recovery tank option available
  • Ready to use - filters and tool kit included
  • Four-wheel cart for easy maneuverability
  • Built-in storage tray with two large 170 mm (6.75") rear wheels and two 60 mm (2.25") front wheels
  • Optional 50' grounded power cord available
HEPA filter with an efficiency of 99.995% at 0.3 micron. Tested IEST-RP-CC001. H14 by MPPS method as per EN 1822 and OSHA compliant. All of our HEPA vacuum systems are aerosol leak tested before leaving our facility - included.
Suction inlet: 1.5"
Ordering information: For available options or accessories; contact your Avantor representative.