Vacuum Cleaner, Dry Recovery-Only, AS-5 HEPA
Supplier: Tiger-Vac International
110750A
77008-222EA 603.39 USD
77008-222
Vacuum Cleaner, Dry Recovery-Only, AS-5 HEPA
Vacuum Cleaners
HEPA filtered vacuum designed for the recovery of asbestos, lead, mold, etc.
  • Designed for the recovery of asbestos, lead, mold, etc.
  • Lightweight and portable
  • Thru-Flow heavy duty motor
  • Four-stage filtration system
  • For dry recovery only
  • Heavy duty plastic recovery tank
  • Ready to use - filters and tool kit included
  • Mounted on a four-wheel dolly for easy maneuverability
HEPA filter with an efficiency of 99.995% at 0.3 micron. Tested IEST-RP-CC001. H14 by MPPS method as per EN 1822 and OSHA compliant. All of our HEPA vacuum systems are aerosol leak tested before leaving our facility - included.
Suction inlet: 1.5"
Certifications: Standard(s) for Electric Vacuum Cleaning Machines and Blower Cleaners (UL-1017).
Ordering information: For available options or accessories; contact your Avantor representative.