Vacuum Cleaner, Dry Recovery-Only, EMI-CWR
Supplier: Tiger-Vac International
110110A
77008-220EA 2830.67 USD
77008-220
Vacuum Cleaner, Dry Recovery-Only, EMI-CWR
Vacuum Cleaners
ESD safe and RFI shielded cleanroom vacuum for dry recovery only. The EMI-CWR comes with a complete toolkit included.
  • Cleanroom compatible with ISO Class 4 (former Class 10); can be used in the pharmaceutical industry
  • Static free and ESD safe
  • EMI/RFI Shielded
  • TELECOM package available
  • Designed for use in contamination controlled environments
  • Powerful dual speed Thru-Flow motor
  • For dry recovery only
  • Made entirely of chrome-polished steel
  • Mounted on a four-wheel dolly for easy maneuverability
  • Includes a complete tool kit
ULPA filter with an efficiency of 99.999% at 0.12 micron. Filtration efficiency of 99.9995% at 0.18 micron. Tested IEST-RP-CC001. U15 by MPPS method as per EN 1822. All of our ULPA vacuum systems are aerosol leak tested before leaving our facility - included.
4-stage filtration system includes an ULPA Filter (U15), double cloth filter, and disposable filter bags. An internal OSHA compliant HEPA filter (H14) is available as a 5th stage of filtration.
Suction inlet: 1.5"
Certifications: ANSI/UL 1017; CSA C22.2 No.243.
Ordering information: For available options or accessories; contact your Avantor representative.