Virtu-Clean® Disposable Cleaning Pads with Scrubber
Supplier: High-Tech Conversions
VCDCS
77008-034CS 200.25 USD
77008-034
Virtu-Clean® Disposable Cleaning Pads with Scrubber
Mops
Not all cleaning pads are equal. Absorbency rates, release rates, coverage, and linting can vary greatly, significantly impacting the finished product.
  • Cleanroom class: ISO 5 (Class 100)
  • Scrubber pad option is designed for more aggressive cleaning and stain removal
  • Holds 7X its weight in cleaning fluid
  • Charge-once-and-clean coverage area of 240 to 300 sq. ft.
  • No Quat binding
  • Compatible with chlorine bleach, hydrogen peroxide, and common disinfectants
Virtu-Clean™ Disposable Cleaning Pads are specially engineered for critical environments, providing maximum coverage, cleaning, and convenience. These disposable, single-use mop pads check all the right boxes – they hold more liquid, release that liquid at a controlled rate over a larger surface area, and reduce friction for more effort-free cleaning.