ULTIMATE 70® Sealed Edge Polyester Cleanroom Wipes
Supplier: High-Tech Conversions
ULT70-1212
77008-010CS 627.28 USD
77008-010
ULTIMATE 70® Sealed Edge Polyester Cleanroom Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
ULTIMATE 70® polyester sealed edge cleanroom wipes are designed for ultra-critical cleanrooms (ISO 3, Class 1 and higher).
  • 100% virgin polyester fibers are continuous with no loose ends
  • Laser cut edges
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Cleanroom laundered and individially inspected
  • Double bagged, vacuum cleanroom packed
  • Autoclavable
  • Meets USP <797> and USP <800> wipe requirements
ULTIMATE 70® wipes are absorbent, heavy-weight, abrasion-resistant, and manufactured from 100% no-run polyester. Individually laser cut, heat sealed-edges eliminate loose fibers.