ProWorks® Catch® Black Nitrile Powder-Free Gloves
Supplier: High-Tech Conversions
GL-NT107BKFL GL-NT107BKFM GL-NT107BKFX GL-NT107BKFXX
77007-974CS 187.73 USD
77007-974 77008-024 77007-874 77007-970
ProWorks® Catch® Black Nitrile Powder-Free Gloves
Gloves
ProWorks® Catch® Black nitrile powder-free disposable gloves feature a heavy pyramid Grip® textured surface that provides an impeccable grip.
  • Industrial grade nitrile construction
  • Powder-free and latex-free
  • 9 mil thickness
  • Ambidextrous
The extra thickness makes these gloves more durable when working on 'rougher' jobs, while keeping hands safe and protected against harsh chemicals, solvents, and paints.
Certifications: Complies with FDA CFR Title 21 Indirect Food Additive Regulations Part 177.
Packaging: Gloves are packaged 100 pieces per box.