Pharma-Glove™ Nitrile Gloves, Sterile, Chemo Approved
Supplier: High-Tech Conversions
PGNS12-2XL PGNS12-3XL PGNS12-LG PGNS12-MD PGNS12-SM PGNS12-XL PGNS12-XS
77008-038CS 521.77 USD
77008-038 77007-842 77007-948 77007-858 77007-952 77007-936 77007-896
Pharma-Glove™ Nitrile Gloves, Sterile, Chemo Approved
Gloves
The Pharma-Glove™ is an ambidextrous glove that is both latex-free and powder-free, with a 12" cuff. This glove is manufactured specifically for use in pharmacies.
  • Sterile nitrile exam gloves
  • 12" length
Pharma-Gloves™ are validated sterile and individually pair packaged with paperless packaging.
Certifications: Chemo approved, ASTM tested gloves.
Packaging: Gloves are packaged 50 pair/bag, 4 bags/pack, 200/case.