VPF Translucent Vinyl Powder-Free Examination Gloves
Supplier: High-Tech Conversions
VPF232 VPF235 VPF236 VPF238
77008-064CS 53.23 USD
77008-064 77007-900 77007-894 77008-008
VPF Translucent Vinyl Powder-Free Examination Gloves
Gloves
VPF Translucent Vinyl Powder-Free Exam Gloves provide a latex-free, powder-free, and economical solution to hand protection.
  • Formulated from a special synthetic component and soft PVC material
  • Gloves are softer and fit snugly and comfortably to hands
Gloves are 100% rubber latex-free, eliminating the risk of latex protein related allergies.
Packaging: Gloves are packaged 100 pieces per box (90 pieces per box for size XL).