Night Angel® Black Nitrile Powder-Free Examination Gloves
Supplier: High-Tech Conversions
NGL222 NGL225 NGL226 NGL228
77007-910CS 75.1 USD
77007-910 77007-872 77007-840 77007-830
Night Angel® Black Nitrile Powder-Free Examination Gloves
Gloves
Night Angel® Black Nitrile Powder-Free Exam Gloves simulate the fit and feel of latex gloves, but are 100% rubber latex-free.
  • 100% synthetic and latex-free
  • Thinner design for maximum dexterity
  • Strong formulation provides superior puncture and tear resistance
  • Excellent barrier protection
Night Angel® gloves are ultra-soft and super flexible to provide more dexterity and tactile sensitivity.
Certifications: Complies with FDA CFR Title 21 Indirect Food Additive Regulations, Part 177.
Packaging: Gloves are packaged 100 pieces per box (90 pieces per box for size XL).