ProWorks® Black Nitrile Powder-Free Examination Gloves
Supplier: High-Tech Conversions
GL-N145FL GL-N145FM GL-N145FS GL-N145FX
77008-070CS 85.4 USD
77008-070 77007-854 77007-868 77007-990
ProWorks® Black Nitrile Powder-Free Examination Gloves
Gloves
ProWorks® Black Nitrile Powder-Free Exam Gloves are formulated with 100% synthetic nitrile polymer. They are latex-free, powder-free, and odor-free.
  • Textured fingers
  • Beaded cuffs
Although these gloves are thicker, they are still much softer and more elastic than most other brands of nitrile gloves of the same thickness. These gloves provide superior puncture, tear, and chemical resistance when compared to latex gloves. The textured surface provides strong grip for both wet and dry applications.
Certifications: Complies with FDA CFR Title 21 Indirect Food Additive Regulations, Part 177.