Precision® Blue Nitrile Powder-Free Examination Gloves
Supplier: High-Tech Conversions
PCS772 PCS775 PCS776 PCS778
77007-832CS 80.87 USD
77007-832 77007-886 77007-930 77007-844
Precision® Blue Nitrile Powder-Free Examination Gloves
Gloves
Ultra-soft and super stretchy Precision® Blue Nitrile Powder-Free Exam Gloves feature a unique low modulus formulation that reduces hand fatigue and provides maximum comfort.
  • 100% synthetic and latex-free
  • Thinner design for maximum dexterity
  • Strong formulation provides superior puncture and tear resistance
  • Excellent barrier protection
Gloves are 100% synthetic and latex-free, avoiding potential allergic reactions to latex proteins. Precision® Nitrile Gloves are made thinner for maximum dexterity, yet they maintain strength to provide superior puncture and tear resistance. These gloves offer excellent barrier protection.
Packaging: Gloves are packaged 100 pieces per box (90 pieces per box for size XL).