Lighthouse Battery Foam Unit with Stainless Steel Ball Valve, 10 Gallon
Supplier: Unimed-Midwest
FPFU516SBV
77007-820EA 3963.42 USD
77007-820
Lighthouse Battery Foam Unit with Stainless Steel Ball Valve, 10 Gallon
Spray Guns
Made for large coverage areas, this unit holds up to 12.5 gallons of fluid and boasts an output distance of 15 to 20 feet.
  • Draws from pre-mixed solution
  • Battery operated
  • Up to 45 minutes of continuous use from one charge
  • Stainless ball valve with foam fan tip
  • Hinged back plate provides tool-free access to components
  • Natural color tank for easy visibility of product levels
  • Hinged lid
  • Integrated hose wraps
Compatible with Peroxigard® RTU (Ready To Use) and concentrate from lighthouse life sciences.
Color-coding options include 5 lid colors (black, blue, green, yellow, red), and 4 discharge hose colors (blue, green, yellow, red).