Hydro AccuMax Dilution System, 1-Button Dilution, Low Flow
Supplier: Unimed-Midwest
HDSC412301
77007-818EA 332.32 USD
77007-818
Hydro AccuMax Dilution System, 1-Button Dilution, Low Flow
Spray Guns
Systems using AccuPro pressure regulation technology eliminate dilution variance by controlling the water pressure into the unit, never allowing the dilution to 'lean out' the desired dilution target.
  • 1-button dilution: low flow bottle fill (1.0 gpm)
  • New, expansive workspace to easily route tubes
  • Easy, tool-free maintenance
  • E-gap and AirGap options to satisfy regulatory standards
  • Same mounting brackets as 'Hydro Systems' AccuPro and AccuDose
High water pressure results in improperly diluted chemicals, poor cleaning performance, and lost chemical sales. AccuPro technology delivers accurate dilution regardless of fluctuations in water pressure.