Lighthouse Foam Unit with Heavy-Duty Trigger Gun, 1.3 Gallon
Supplier: Unimed-Midwest
FPUF516PSG
77007-810EA 304.29 USD
77007-810
Lighthouse Foam Unit with Heavy-Duty Trigger Gun, 1.3 Gallon
Spray Guns
This compact unit features a manual pump and is ideal for small jobs and spot cleaning.
  • Maximum portability for small jobs and spot cleaning
  • Pressure relief valve for safety
  • Viton seals for chemical compatibility
  • Natural color tank for easy visibility of product levels
  • Product level fill markings in gallons and liters
  • Extra-wide stability base
This compact unit with manual pump features a heavy-duty trigger gun for additional durability.