Lighthouse Pail Hand Pump for Five-Gallon Pails
Supplier: Unimed-Midwest
DPHP511505
77007-800EA 96.18 USD
77007-800
Lighthouse Pail Hand Pump for Five-Gallon Pails
Pumps Drum/Transfer Pumps
The MR50-5 plastic pail hand pump is a compact hand pump for 5-gallon buckets that features a polyethylene body and Viton® O-rings.
  • Can be used for detergents and mild chemicals from 5 gallon pails
  • Includes rigid discharge spout, inlet tubes and adapters will fit ¾" drum fittings, 2" bung fittings, flex spouts and rigid containers
  • Adjustable output from 2, 4, 6 or 8 oz. per stroke
This poly hand pump is specifically designed for use with 5-gallon pails and can be used for dispensing detergents and mild chemicals. This 5-gallon bucket pump includes a rigid discharge spout, inlet tubes, and adapters which will fit ¾" drum fittings, 2" bung fittings, flex spouts, and rigid containers. This hand-operated 5-gallon transfer pump can be adjusted to output 2, 4, 6 or 8 oz. per stroke.