Anti-AAV Rabbit Monoclonal Antibody [clone: 24F5]

Supplier: Genscript
A02207-40
77007-634EA 362.41 USD
77007-634
Anti-AAV Rabbit Monoclonal Antibody [clone: 24F5]
Antibodies
Adeno-associated virus (AAV) is a single-stranded DNA virus that commonly infects humans without pathology. Therefore, AAV can be engineered to deliver DNA to target cells as a vector for gene therapy. There are several serotypes including AAV type 1, 2, 5, 6, 8, 9, DJ and rh.1. Among them, AAV2 is the best characterized and most commonly used. Cap gene of AAV gives rise to three capsid proteins, VP1, VP2 and VP3, with molecular weight of 87, 72 and 62 kDa respectively. These capsid proteins assemble into a near-spherical protein shell of 60 subunits for AAV.

  • High specificity
  • High affinity
  • High stability

GenScript's AAV antibody is a useful tool for evaluation of virus expression level, quantification of virus titer, identification of capsid proteins in AAV production, and analysis of AAV immunogenicity.

Type:
Antigen: AAV
Clonality: Monoclonal
Clone: 24F5
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR