Anti-GST Tag Rabbit Polyclonal Antibody

Supplier: Genscript
A00097-40
77007-622EA 104.04 USD
77007-622
Anti-GST Tag Rabbit Polyclonal Antibody
Antibodies
GenScript Rabbit Anti-GST-tag Polyclonal Antibody is highly purified from rabbit antiserum by immunoaffinity chromatography. The immunogen used is a recombinant version of glutathione-S-transferase from Schistosoma japonicum. Glutathione-S-transferase is an epitope tag in generating fusion proteins. Antibodies specific to fusion proteins are useful in monitoring protein expression in Western Blots as well as immunoaffinity purification of proteins expressed in E. coli. The glutathione-S-transferases are a family of enzymes that catalyze the addition of the tripeptide glutathione to endogenous and xenobiotic substrates which have electrophilic functional groups. They play an important role in the detoxication and metabolism of many xenobiotic and endobiotic compounds. The glutathione adducts produced have increased solubility in water and are subsequently enzymatically degraded to mercapturates and excreted.

  • High specificity
  • High affinity
  • High stability

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: GST Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. japonicum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR