Anti-Protein A Mouse Monoclonal Antibody (Biotin) [clone: 5G10A7]

Supplier: Genscript
A01779-40
77007-572EA 112.3 USD
77007-572
Anti-Protein A Mouse Monoclonal Antibody (Biotin) [clone: 5G10A7]
Antibodies
Protein A, a bacterial cell wall protein isolated from Staphylococcus aureus, binds to mammalian IgGs mainly through Fc regions. The protein A is most commonly bound to a chromatography resin, used for production of antibody drugs or reagents. Protein A antibody can be used for quantification of protein A in sample foods or for evaluation of protein A resin. This protein A mAb is advantageous due to its IgG1 isotype, for mouse IgG1 has weak binding capacity to protein A.GenScript protein A Antibody [biotin], mAb, mouse is protein A antibody conjugated with biotin. The conjugated antibody is suitable for detecting the expression level of protein A.

  • High specificity
  • High affinity
  • High stability

GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.

Type: Primary
Antigen: Protein A
Clonality: Monoclonal
Clone: 5G10A7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR