Anti-VHH Rabbit Monoclonal Antibody (iFluor 647) [clone: 45H8]

Supplier: Genscript
A02170-200
77007-504EA 249.18 USD
77007-504
Anti-VHH Rabbit Monoclonal Antibody (iFluor 647) [clone: 45H8]
Antibodies
A camelid single-domain antibody (sdAb, called Nanobody by Ablynx, the developer) is a peptide chain of about 110 amino acids long, comprising one variable domain of a heavy-chain antibody, these are called VHH fragments. Like a whole antibody, it is able to bind selectively to a specific antigen. Single-domain antibodies allow a broad range of applications in biotechnical as well as therapeutic uses due to their small size, simple production, and high affinity. However, their sequences may increase the risk of immunogenicity and anti-drug antibody (ADA) development in humans, and thus, sdAbs are routinely humanized during development.

  • MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
  • No cross reactivity with other species
  • Specific to camelid IgG2 and IgG3
  • Recognize conformational epitopes of the VHH domain
  • Recognize both camelid and humanized VHHs

GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.

Type:
Antigen: VHH
Clonality: Monoclonal
Clone: 45H8
Conjugation: iFluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR