Anti-Bevacizumab Mouse Monoclonal Antibody [clone: 11C1]

Supplier: Genscript
A02185-40
77007-476EA 153.9 USD
77007-476
Anti-Bevacizumab Mouse Monoclonal Antibody [clone: 11C1]
Antibodies
Bevacizumab (Avastin) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of a number of types of cancers and a specific eye disease. Bevacizumab binds to a continuous epitope in VEGF, preventing interaction of VEGF with VEGFR on the surface of endothelial cells. It can be used to inhibit solid tumours growth and metastasis by weakening tumour angiogenesis.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Bevacizumab
Clonality: Monoclonal
Clone: 11C1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR