Methyl-4-bromocrotonate 80%

Supplier: Ambeed
Danger

A787584-10G A787584-1G A787584-25G A787584-5G
77006-470EA 132 USD
77006-470 77006-472 77006-474 77006-476 77006-478
Methyl-4-bromocrotonate 80%
Methyl 4-bromocrotonate

Caution: for Research Use Only

Formula: C₅H₇BrO₂
MDL Number: MFCD00000246
CAS Number: 1117-71-1
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR