4-Ethylbenzenesulfonyl chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Ethylbenzene-1-sulfonyl chloride, 4-Ethylbenzene-1-sulphonyl chloride
A754699-1G
A754699-25G
A754699-5G
77006-270EA
7
USD
77006-270
77006-272
77006-274
4-Ethylbenzenesulfonyl chloride 98%
4-Ethylbenzenesulphonyl chloride
Caution: for Research Use Only
Formula:
C₈H₉ClO₂S |
MDL Number:
MFCD00041480 CAS Number: 16712-69-9 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...