3,4,5-Trifluoroanisole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2,3-Trifluoro-5-methoxybenzene, Trifluoro-3,4,5-anisole, 1-Methoxy-3,4,5-trifluorobenzene
A691792-1G
A691792-25G
A691792-5G
77005-930EA
21
USD
77005-930
77005-932
77005-934
3,4,5-Trifluoroanisole 97%
3,4,5-Trifluoroanisole
Caution: for Research Use Only
Formula:
C₇H₅F₃O MW: 162.11 |
MDL Number:
MFCD00042397 CAS Number: 203245-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...