(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(+)-MOP, (R)-(2'-Methoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine, (R)-(+)-2-(Diphenylphosphino)-2'-methoxy-1,1'-binaphthyl, (R)-MOP

A666526-100MG A666526-1G A666526-250MG
77005-772EA 21 USD
77005-772 77005-774 77005-776
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl 98%
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl

Caution: for Research Use Only

Formula: C₃₃H₂₅OP
MDL Number: MFCD00269686
CAS Number: 145964-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR