5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dimethoxybenzo[b]thiophene-2-carboxylic acid

A626637-100MG A626637-1G A626637-250MG
77005-556EA 17 USD
77005-556 77005-558 77005-560
5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid 95%
5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid

Caution: for Research Use Only

Formula: C₁₁H₁₀O₄S
MDL Number: MFCD08275132
CAS Number: 23046-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR