Iminodiacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,2'-Azanediyldiacetic acid

A617225-25G A617225-1000G A617225-100G A617225-500G
76844-370EA 9 USD
76844-370 77005-494 77005-496 77005-498
Iminodiacetic acid 98%
Iminodiacetic acid

Caution: for Research Use Only

Formula: C₄H₇NO₄
MDL Number: MFCD00004280
CAS Number: 142-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR