4-tert-Butylbenzenesulfonyl chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride
A392201-10G
A392201-25G
A392201-5G
77003-764EA
10
USD
77003-764
77003-766
77003-768
4-tert-Butylbenzenesulfonyl chloride 98%
4-tert-Butylbenzenesulphonyl chloride
Caution: for Research Use Only
Formula:
C₁₀H₁₃ClO₂S MW: 232.73 Boiling Pt: 165 Melting Pt: 83 |
MDL Number:
MFCD00007449 CAS Number: 15084-51-2 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...