Ferrocenecarbaldehyde 97%

Supplier: Ambeed
Danger

A390081-10G A390081-1G A390081-25G A390081-5G
77003-744EA 28 USD
77003-744 77003-746 77003-748 77003-750
Ferrocenecarbaldehyde 97%
Ferrocenecarbaldehyde

Caution: for Research Use Only

Formula: C₁₁H₁₀FeO
MDL Number: MFCD00001429
CAS Number: 12093-10-6
UN: 3467
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR