3-Iodo-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Iodopyrazole

A362440-1G A362440-25G A362440-5G A362440-10G A362440-250MG
103826-318EA 7 USD
103826-318 103837-334 76840-548 76840-550 76840-552 77003-374 77003-376
3-Iodo-1H-pyrazole 98%
3-Iodo-1H-pyrazole

Caution: for Research Use Only

Formula: C₃H₃IN₂
MW: 193.98 g/mol
Boiling Pt: 291.9 °C (760 mm Hg)
Melting Pt: 74…75 °C
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS Number: 4522-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR