α-Naphtholphthalein 98%

Supplier: Ambeed
Warning

Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

A352623-100G A352623-25G A352623-5G A352623-1G
77003-260EA 268 USD
77003-260 77003-262 77003-264 77881-965
α-Naphtholphthalein 98%
α-Naphtholphthalein

Caution: for Research Use Only

Formula: C₂₈H₁₈O₄
MW: 418.45 g/mol
MDL Number: MFCD00036202
CAS Number: 596-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR