1-Cyclohexene-1-acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclohexenylacetic acid

A341826-10G A341826-1G A341826-250MG A341826-25G A341826-5G
77003-146EA 81 USD
77003-146 77003-148 77003-150 77003-152 77003-154
1-Cyclohexene-1-acetic acid 97%
1-Cyclohexene-1-acetic acid

Caution: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00015482
CAS Number: 18294-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR