1-Cyclohexene-1-acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Cyclohexenylacetic acid
A341826-10G
A341826-1G
A341826-250MG
A341826-25G
A341826-5G
77003-146EA
81
USD
77003-146
77003-148
77003-150
77003-152
77003-154
1-Cyclohexene-1-acetic acid 97%
1-Cyclohexene-1-acetic acid
Caution: for Research Use Only
Formula:
C₈H₁₂O₂ |
MDL Number:
MFCD00015482 CAS Number: 18294-87-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...