Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 97%

Supplier: Ambeed
Warning

Synonyms: Dibenzo-18-crown-6

A325441-100G A325441-10G A325441-1G A325441-25G A325441-5G A325441-500G
77002-918EA 56 USD
77002-918 77002-920 77002-922 77002-924 77002-926 77881-910
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 97%
Dibenzo-18-crown-6 (Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin (II) ionophore I)

Caution: for Research Use Only

Formula: C₂₀H₂₄O₆
MDL Number: MFCD00005098
CAS Number: 14187-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR