Gallic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,4,5-Trihydroxybenzoic acid
A311911-1000G
A311911-100G
A311911-25G
A311911-500G
A311911-50MG
77002-698EA
49
USD
77002-698
77002-700
77002-702
77002-704
77002-706
77672-594
Gallic acid 98%
Gallic acid
Caution: for Research Use Only
Formula:
C₇H₆O₅ MW: 170.12 g/mol Boiling Pt: 501 |
MDL Number:
MFCD00002510 CAS Number: 149-91-7 Merck Index: 13,04366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...