Gallic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid

A311911-1000G A311911-100G A311911-25G A311911-500G A311911-50MG
77002-698EA 49 USD
77002-698 77002-700 77002-702 77002-704 77002-706 77672-594
Gallic acid 98%
Gallic acid

Caution: for Research Use Only

Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
MDL Number: MFCD00002510
CAS Number: 149-91-7
Merck Index: 13,04366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR