DNase I Set
Supplier: Zymo Research
E1010
77001-900EA 133.65 USD
77001-900
DNase I Set
Enzymes
Zymo research DNase I is a recombinant endonuclease that cleaves single- and double-stranded DNA. It digests DNA more efficiently than other DNase I, making it optimal for sensitive applications like NGS (RNA-seq) and PCR. It is extensively purified, and quality controlled to ensure RNase-free and protease-free.
  • Lyophilized enzyme
  • Used for selectively degrading DNA in the presence of RNA
DNase is suited for applications such as nick translation, production of random fragments, cleavage of genomic DNA for footprinting, removal of DNA template after in vitro transcription and removal of DNA from RNA samples prior to applications such as RT-PCR. It is compatible with all of our RNA kits featuring in-column DNase digestion.
Zymo Research DNase I is lyophilized and supplied with DNA digestion buffer. It dissolves in water within seconds and remains stable for up to 10 freeze-thaw cycles.
Ordering information:
Customized, OEM, and bulk orders are available on request.
Delivery information: The set supplied with DNA digestion buffer (4 ml).