Microbial Community DNA Standard, ZymoBIOMICS®, Zymo Research
Supplier: Zymo Research
D6306
77001-846EA 245.16 USD
77001-846
Microbial Community DNA Standard, ZymoBIOMICS®, Zymo Research
Standards Biological Standards
The ZymoBIOMICS Microbial Community DNA Standard is designed to assess bias, errors and other artifacts after the step of nucleic acid purification.
  • Accurate composition: Composition cross-validated with multiple types of measurements.
  • Negligible impurity: Guaranteed to contain < 0.01% foreign microbial DNA.
  • Wide range of GC content: 15%-85%, for assessing bias caused by GC content variation.
Theoretical Composition based on Genomic DNA are Listeria monocytogenes - 12%, Pseudomonas aeruginosa - 12%, Bacillus subtilis - 12%, Escherichia coli - 12%, Salmonella enterica - 12%, Lactobacillus fermentum - 12%, Enterococcus faecalis - 12%, Staphylococcus aureus - 12%, Saccharomyces cerevisiae - 2%, and Cryptococcus neoformans - 2%.