Tris(carbobenzoxy)-L-arginine 98%

Supplier: Ambeed
Warning

Synonyms: Nα,Nδ,Nomega-Tris(benzyloxycarbonyl)-L-arginine, Tri-Cbz-L-arginine, Z-Arg(Z)2-OH, Z-Arg(Cbz)₂-OH

A272674-1G A272674-5G A272674-250MG
77001-616EA 13 USD
77001-616 77001-618 77601-170
Tris(carbobenzoxy)-L-arginine 98%
Tris(carbobenzoxy)-L-arginine

Caution: for Research Use Only

Formula: C₃₀H₃₂N₄O₈
MW: 576.61 g/mol
MDL Number: MFCD00038693
CAS Number: 14611-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR