(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 98%

Supplier: Ambeed
Warning

A272063-100G A272063-10G A272063-250MG
103813-082EA 11 USD
103813-082 103852-994 77001-598 77001-600 77001-602
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene 98%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene

Caution: for Research Use Only

Formula: C₂₀H₁₆N₂
Boiling Pt: 480.8 °C (760 mm Hg)
Melting Pt: 242…244 °C
Flash Pt: 293.854 °C
Storage Temperature: Ambient
MDL Number: MFCD00145204
CAS Number: 18741-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR