2-Methoxy-3-nitropyridine 97%
Supplier: Ambeed
|
Warning
|
A260561-100G
A260561-25G
A260561-5G
77000-924EA
91
USD
77000-924
77000-926
77000-928
2-Methoxy-3-nitropyridine 97%
2-Methoxy-3-nitropyridine
Caution: for Research Use Only
Formula:
C₆H₆N₂O₃ |
MDL Number:
MFCD00023459 CAS Number: 20265-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...