D-(-)-N-Ethylglucamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Ethyl-D-glucamine, (-)-1-Deoxy-1-(ethylamino)-D-glucitol, (-)-1-Ethylamino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol, (-)-N-Ethyl-D-glucamine
A219049-1000G
A219049-100G
A219049-25G
A219049-500G
76999-854EA
99
USD
76999-854
76999-856
76999-858
76999-860
D-(-)-N-Ethylglucamine 98%
D(-)-N-Ethylglucamine
Caution: for Research Use Only
Formula:
H₃CCH₂NHCH₂(CH(OH))₄CH₂OH |
MDL Number:
MFCD03789564 CAS Number: 14216-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...