D-(-)-N-Ethylglucamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Ethyl-D-glucamine, (-)-1-Deoxy-1-(ethylamino)-D-glucitol, (-)-1-Ethylamino-1-deoxy-D-glucitol, (2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol, (-)-N-Ethyl-D-glucamine

A219049-1000G A219049-100G A219049-25G A219049-500G
76999-854EA 99 USD
76999-854 76999-856 76999-858 76999-860
D-(-)-N-Ethylglucamine 98%
D(-)-N-Ethylglucamine

Caution: for Research Use Only

Formula: H₃CCH₂NHCH₂(CH(OH))₄CH₂OH
MDL Number: MFCD03789564
CAS Number: 14216-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR