L-Aspartic acid 4-cyclohexyl ester 98%

Supplier: Ambeed
Warning

Synonyms: H-Asp(OcHex)-OH hydrochloride salt, H-Asp(OcHx)-OH, H-Asp(OcHex)-OH

A213441-1G A213441-250MG A213441-5G
76999-670EA 10 USD
76999-670 76999-672 76999-674
L-Aspartic acid 4-cyclohexyl ester 98%
L-Aspartic acid 4-cyclohexyl ester

Caution: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.249 g/mol
MDL Number: MFCD00190762
CAS Number: 112259-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR