Tetrakis(triphenylphosphine)palladium(0) 98%

Supplier: Ambeed
Warning

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

A211391-10G A211391-25G A211391-5G A211391-100G
103818-106EA 34 USD
103818-106 103848-800 76999-618 77087-206 77087-208 77881-508
Tetrakis(triphenylphosphine)palladium(0) 98%
Tetrakis(triphenylphosphine)palladium (0)

Caution: for Research Use Only

Formula: C₇₂H₆₀P₄Pd
Melting Pt: 100…105 °C
Flash Pt: 181.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00010012
CAS Number: 14221-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR