Sirtinol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(((2-Hydroxynaphthalen-1-yl)methylene)amino)-N-(1-phenylethyl)benzamide
A204147-10MG
A204147-1MG
A204147-50MG
A204147-5MG
76999-442EA
81.99
USD
76999-442
76999-444
76999-446
76999-448
Sirtinol 98%
Sirtinol
Caution: for Research Use Only
Formula:
C₂₆H₂₂N₂O₂ |
MDL Number:
MFCD00810186 CAS Number: 410536-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...