Sirtinol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(((2-Hydroxynaphthalen-1-yl)methylene)amino)-N-(1-phenylethyl)benzamide

A204147-10MG A204147-1MG A204147-50MG A204147-5MG
76999-442EA 81.99 USD
76999-442 76999-444 76999-446 76999-448
Sirtinol 98%
Sirtinol

Caution: for Research Use Only

Formula: C₂₆H₂₂N₂O₂
MDL Number: MFCD00810186
CAS Number: 410536-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR