(R)-Methyl-2-amino-2-phenylacetate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-(-)-2-Phenylglycine methyll ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, Methyl (R)-(-)-2-Amino-2-phenylacetate hydrochloride, H-D-Phg-OMe·HCl, H-D-Phg-OMe.HCl, (R)-(-)-2-Amino-2-phenylacetic acid methyll ester hydrochloride
A189810-100G
A189810-25G
A189810-500G
A189810-5G
76998-924EA
45
USD
76998-924
76998-926
76998-928
76998-930
(R)-Methyl-2-amino-2-phenylacetate hydrochloride 98%
(R)-Methyl-2-amino-2-phenylacetate hydrochloride
Caution: for Research Use Only
Formula:
C₉H₁₁NO₂·HCl MW: 201.7 g/mol |
MDL Number:
MFCD00137487 CAS Number: 19883-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...