(R)-Methyl-2-amino-2-phenylacetate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: D-(-)-2-Phenylglycine methyll ester hydrochloride, (R)-2-Phenylglycine methyl ester hydrochloride, Methyl (R)-(-)-2-Amino-2-phenylacetate hydrochloride, H-D-Phg-OMe·HCl, H-D-Phg-OMe.HCl, (R)-(-)-2-Amino-2-phenylacetic acid methyll ester hydrochloride

A189810-100G A189810-25G A189810-500G A189810-5G
76998-924EA 45 USD
76998-924 76998-926 76998-928 76998-930
(R)-Methyl-2-amino-2-phenylacetate hydrochloride 98%
(R)-Methyl-2-amino-2-phenylacetate hydrochloride

Caution: for Research Use Only

Formula: C₉H₁₁NO₂·HCl
MW: 201.7 g/mol
MDL Number: MFCD00137487
CAS Number: 19883-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR